Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YPT53
Entrez Gene ID
855631
Gene Name
Gene Aliases
YNL093W
Chromosome Location
Chr.XIV: 449868 - 450530 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182931.3 NP_014306.3 1 - 1 548 105

View Full Product Details