Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEU4
Entrez Gene ID
855619
Gene Name
Gene Aliases
YNL104C
Chromosome Location
Chr.XIV: 424895 - 426754 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182942.1 NP_014295.1 1 - 1 1748 93

View Full Product Details