Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ESBP6
Entrez Gene ID
855598
Gene Name
Gene Aliases
MCH3, YNL125C
Chromosome Location
Chr.XIV: 390146 - 392167 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182963.1 NP_014274.1 1 - 1 1655 74

View Full Product Details