Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CBK1
Entrez Gene ID
855561
Gene Name
Gene Aliases
YNL161W
Chromosome Location
Chr.XIV: 332597 - 334867 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182999.3 NP_014238.3 1 - 1 2155 65

View Full Product Details