Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRG9
Entrez Gene ID
855508
Gene Name
Gene Aliases
YNL213C
Chromosome Location
Chr.XIV: 246460 - 247104 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183051.1 NP_014186.1 1 - 1 550 78

View Full Product Details