Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PFS2
Entrez Gene ID
855399
Gene Name
Gene Aliases
YNL317W
Chromosome Location
Chr.XIV: 40619 - 42016 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183155.1 NP_014082.1 1 - 1 1255 80

View Full Product Details