Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EGT2
Entrez Gene ID
855389
Gene Name
Gene Aliases
YNL327W
Chromosome Location
Chr.XIV: 24048 - 27173 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183165.2 NP_014072.2 1 - 1 3048 90

View Full Product Details