Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SOL1
Entrez Gene ID
855769
Gene Name
Gene Aliases
YNR034W
Chromosome Location
Chr.XIV: 690321 - 691286 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183211.3 NP_014432.3 1 - 1 750 74

View Full Product Details