Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSO1
Entrez Gene ID
855785
Gene Name
Gene Aliases
YNR049C
Chromosome Location
Chr.XIV: 713023 - 713655 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183226.3 NP_014447.3 1 - 1 550 95

View Full Product Details