Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YAP7
Entrez Gene ID
854129
Gene Name
Gene Aliases
YOL028C
Chromosome Location
Chr.XV: 270633 - 271370 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183282.1 NP_014614.1 1 - 1 650 84

View Full Product Details