Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GSH2
Entrez Gene ID
854108
Gene Name
Gene Aliases
YOL049W
Chromosome Location
Chr.XV: 238619 - 240094 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183303.1 NP_014593.1 1 - 1 1344 81

View Full Product Details