Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INP54
Entrez Gene ID
854089
Gene Name
Gene Aliases
YOL065C
Chromosome Location
Chr.XV: 204731 - 205885 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183320.1 NP_014576.1 1 - 1 1051 58

View Full Product Details