Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPM2
Entrez Gene ID
854024
Gene Name
Gene Aliases
TYW4, YOL141W
Chromosome Location
Chr.XV: 56452 - 58539 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183395.2 NP_014500.2 1 - 1 1856 117

View Full Product Details