Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSF3
Entrez Gene ID
854019
Gene Name
Gene Aliases
YOL146W
Chromosome Location
Chr.XV: 48864 - 49448 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183400.1 NP_014495.2 1 - 1 345 86

View Full Product Details