Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RSB1
Entrez Gene ID
854214
Gene Name
Gene Aliases
YOR049C
Chromosome Location
Chr.XV: 422668 - 423732 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183468.1 NP_014692.1 1 - 1 852 60

View Full Product Details