Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS5
Entrez Gene ID
854235
Gene Name
Gene Aliases
GRD2, PEP10, VPT5, YOR069W
Chromosome Location
Chr.XV: 453768 - 455795 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183488.1 NP_014712.2 1 - 1 1646 86

View Full Product Details