Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NRT1
Entrez Gene ID
854237
Gene Name
Gene Aliases
THI71, YOR071C, YOR29-22
Chromosome Location
Chr.XV: 459480 - 461276 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183490.1 NP_014714.1 1 - 1 248 81

View Full Product Details