Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RKI1
Entrez Gene ID
854262
Gene Name
Gene Aliases
YOR095C
Chromosome Location
Chr.XV: 503552 - 504328 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183514.1 NP_014738.1 1 - 1 651 106

View Full Product Details