Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PIN2
- Entrez Gene ID
-
854271
- Gene Name
- Gene Aliases
-
YOR104W
- Chromosome Location
-
Chr.XV: 517642 - 518490 on Build R64
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001183523.1 |
NP_014747.1 |
1 - 1 |
644 |
91 |
- Gene Symbol
- 854272
- Entrez Gene ID
-
854272
- Gene Name
- Gene Aliases
-
YOR105W
- Chromosome Location
-
Chr.XV: 518195 - 518521 on Build R64
View Full Product Details