Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LEU9
Entrez Gene ID
854275
Gene Name
Gene Aliases
YOR108W
Chromosome Location
Chr.XV: 523027 - 524841 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183527.1 NP_014751.1 1 - 1 1751 93

View Full Product Details