Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
INP53
Entrez Gene ID
854276
Gene Name
Gene Aliases
SJL3, SOP2, YOR109W
Chromosome Location
Chr.XV: 525278 - 528601 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183528.3 NP_014752.3 1 - 1 3255 85

View Full Product Details