Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALE1
Entrez Gene ID
854346
Gene Name
Gene Aliases
LCA1, LPT1, SLC4, YOR175C
Chromosome Location
Chr.XV: 659815 - 661674 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183594.1 NP_014818.1 1 - 1 1650 99

View Full Product Details