Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIS3
Entrez Gene ID
854377
Gene Name
Gene Aliases
HIS10, HIS8, YOR202W
Chromosome Location
Chr.XV: 721946 - 722608 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183621.1 NP_014845.1 1 - 1 539 73

View Full Product Details