Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RUD3
Entrez Gene ID
854391
Gene Name
Gene Aliases
GRP1, YOR216C
Chromosome Location
Chr.XV: 747525 - 748979 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183635.3 NP_014859.3 1 - 1 1352 78

View Full Product Details