Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CDC31
Entrez Gene ID
854431
Gene Name
Gene Aliases
DSK1, YOR257W
Chromosome Location
Chr.XV: 811008 - 811493 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183676.3 NP_014900.3 1 - 1 351 89

View Full Product Details