Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPT4
Entrez Gene ID
854433
Gene Name
Gene Aliases
CRL13, PCS1, SUG2, YOR259C
Chromosome Location
Chr.XV: 812395 - 813708 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183678.3 NP_014902.3 1 - 1 1249 87

View Full Product Details