Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GCD1
Entrez Gene ID
854434
Gene Name
Gene Aliases
TRA3, YOR260W
Chromosome Location
Chr.XV: 813984 - 815720 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183679.1 NP_014903.1 1 - 1 1654 91

View Full Product Details