Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GNT1
Entrez Gene ID
854498
Gene Name
Gene Aliases
YOR320C
Chromosome Location
Chr.XV: 913619 - 915094 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183740.3 NP_014965.3 1 - 1 1351 68

View Full Product Details