Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VMA4
Entrez Gene ID
854509
Gene Name
Gene Aliases
YOR332W
Chromosome Location
Chr.XV: 943656 - 944357 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183752.3 NP_014977.3 1 - 1 650 94

View Full Product Details