Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TFC8
Entrez Gene ID
856100
Gene Name
Gene Aliases
YPL007C
Chromosome Location
Chr.XVI: 542082 - 543848 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183821.1 NP_015318.1 1 - 1 1648 106

View Full Product Details