Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTF19
Entrez Gene ID
856089
Gene Name
Gene Aliases
MCM18, YPL018W
Chromosome Location
Chr.XVI: 517651 - 518760 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183832.1 NP_015307.1 1 - 1 1048 73

View Full Product Details