Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BTS1
Entrez Gene ID
856036
Gene Name
Gene Aliases
YPL069C
Chromosome Location
Chr.XVI: 422885 - 423892 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183883.1 NP_015256.1 1 - 1 752 89

View Full Product Details