Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NOG1
Entrez Gene ID
856012
Gene Name
Gene Aliases
YPL093W
Chromosome Location
Chr.XVI: 370978 - 372921 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183907.1 NP_015232.1 1 - 1 1849 102

View Full Product Details