Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BEM3
Entrez Gene ID
855988
Gene Name
Gene Aliases
YPL115C
Chromosome Location
Chr.XVI: 332100 - 335486 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183929.1 NP_015210.1 1 - 1 3248 124

View Full Product Details