Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS30
Entrez Gene ID
855983
Gene Name
Gene Aliases
APG6, ATG6, VPT30, YPL120W
Chromosome Location
Chr.XVI: 322071 - 323744 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183934.1 NP_015205.1 1 - 1 1546 96

View Full Product Details