Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SVS1
Entrez Gene ID
855940
Gene Name
Gene Aliases
YPL163C
Chromosome Location
Chr.XVI: 241918 - 242700 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183977.1 NP_015162.1 1 - 1 641 65

View Full Product Details