Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OYE3
Entrez Gene ID
855932
Gene Name
Gene Aliases
YPL171C, ZRG6
Chromosome Location
Chr.XVI: 226169 - 227371 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183985.1 NP_015154.1 1 - 1 1151 101

View Full Product Details