Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARP7
Entrez Gene ID
856146
Gene Name
Gene Aliases
RSC11, SWP61, YPR034W
Chromosome Location
Chr.XVI: 639525 - 640958 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184131.1 NP_015359.1 1 - 1 1251 76

View Full Product Details