Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ISA2
Entrez Gene ID
856180
Gene Name
Gene Aliases
YPR067W
Chromosome Location
Chr.XVI: 682220 - 682777 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184164.1 NP_015392.1 1 - 1 450 99

View Full Product Details