Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TKL1
Entrez Gene ID
856188
Gene Name
Gene Aliases
YPR074C
Chromosome Location
Chr.XVI: 692796 - 694838 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184171.1 NP_015399.1 1 - 1 1948 66

View Full Product Details