Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSS18
Entrez Gene ID
856253
Gene Name
Gene Aliases
YPR134W
Chromosome Location
Chr.XVI: 798051 - 798857 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184231.1 NP_015460.1 1 - 1 747 88

View Full Product Details