Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUR1
Entrez Gene ID
856281
Gene Name
Gene Aliases
YPR158W
Chromosome Location
Chr.XVI: 843262 - 844020 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184255.1 NP_015484.1 1 - 1 651 73

View Full Product Details