Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GIM4
Entrez Gene ID
856715
Gene Name
Gene Aliases
PFD2, YEL003W
Chromosome Location
Chr.V: 148176 - 148599 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001178818.1 NP_010913.2 2 - 2 147 93

View Full Product Details