Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPT14
Entrez Gene ID
855928
Gene Name
Gene Aliases
CWH6, GPI3, YPL175W
Chromosome Location
Chr.XVI: 218630 - 220088 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183989.1 NP_015150.2 2 - 2 1050 110

View Full Product Details