Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC27
Entrez Gene ID
852740
Gene Name
Gene Aliases
YGL137W
Chromosome Location
Chr.VII: 249869 - 252738 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181002.1 NP_011378.1 2 - 2 2550 108

View Full Product Details