Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRSF9
Entrez Gene ID
100154637
Gene Name
serine and arginine rich splicing factor 9
Gene Aliases
-
Chromosome Location
Chr.14: 40425412 - 40433950 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195566.1 NP_001182495.1 2 - 3 377 69
GenBank mRNA AK234556.1 - 2 - 3 461 69
AK239990.1 - 2 - 3 465 69
AK392301.1 - 2 - 3 459 69
AY610301.1 - 2 - 3 377 69

View Full Product Details