Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC110258517
Entrez Gene ID
110258517
Gene Name
olfactory receptor-like protein OLF4
Gene Aliases
-
Chromosome Location
Chr.Unknown: 269747 - 281199 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF042021.1 - 2 - 2 448 103

View Full Product Details