Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DECR1
Entrez Gene ID
503544
Gene Name
2,4-dienoyl-CoA reductase 1
Gene Aliases
DECR
Chromosome Location
Chr.4: 46731359 - 46767427 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190232.2 NP_001177161.2 1 - 2 83 101
GenBank mRNA AJ301324.2 - 1 - 2 52 101
AK230726.1 - 1 - 2 83 101
EW352113.2 - 1 - 2 96 101
HM004547.2 - 1 - 2 68 101

View Full Product Details