Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HHEX
Entrez Gene ID
397232
Gene Name
hematopoietically expressed homeobox
Gene Aliases
-
Chromosome Location
Chr.14: 104194424 - 104199684 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001244579.1 NP_001231508.1 3 - 4 629 88
GenBank mRNA AJ316616.1 - 3 - 4 493 88

View Full Product Details