Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COL11A1
Entrez Gene ID
397175
Gene Name
collagen type XI alpha 1 chain
Gene Aliases
COLL11A1
Chromosome Location
Chr.4: 115638381 - 115841679 on Build Sscrofa11.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001929372.7 XP_001929407.6 - 2175 120
GenBank mRNA AJ536287.1 - 8 - 9 384 120

View Full Product Details